Product Details

SNP ID
rs200866315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:115122768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTAAACAAAACTTACCACTGCTG[A/G]TGATGGTACATGAATACTAGCAACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRCH2 PubMed Links

Gene Details

Gene
LRCH2
Gene Name
leucine rich repeats and calponin homology domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243963.1 2123 Missense Mutation CCA,TCA P681S NP_001230892.1
NM_020871.3 2123 Missense Mutation CCA,TCA P698S NP_065922.3
XM_006724724.3 2123 Missense Mutation CCA,TCA P691S XP_006724787.2
XM_017029696.1 2123 Intron XP_016885185.1
XM_017029697.1 2123 Intron XP_016885186.1

View Full Product Details