Product Details

SNP ID
rs201154448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:35919994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTACCGCCTCCCGATTACTGTGCA[C/T]AATATTTGCCGCTGGAACCAGAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFAP47 PubMed Links

Gene Details

Gene
CFAP47
Gene Name
cilia and flagella associated protein 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304548.1 254 Silent Mutation CAC,CAT H65H NP_001291477.1
NM_152632.3 254 Silent Mutation CAC,CAT H65H NP_689845.2
XM_017029452.1 254 Silent Mutation CAC,CAT H65H XP_016884941.1
XM_017029453.1 254 Silent Mutation CAC,CAT H65H XP_016884942.1

View Full Product Details