Product Details

SNP ID
rs200894457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70288500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGTCAACTCATTGTTGTAGTCC[A/G]GGTGGTGTACCCTCTGCAAGACACA
Phenotype
MIM: 301770 MIM: 300521 MIM: 300632
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARR3 PubMed Links

Gene Details

Gene
ARR3
Gene Name
arrestin 3
There are no transcripts associated with this gene.

Gene
KIF4A
Gene Name
kinesin family member 4A
There are no transcripts associated with this gene.

Gene
PDZD11
Gene Name
PDZ domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016484.4 534 Missense Mutation CCG,CTG P34L NP_057568.1
XM_011530973.2 534 Missense Mutation CCG,CTG P34L XP_011529275.1
XM_017029568.1 534 Missense Mutation CCG,CTG P66L XP_016885057.1
Gene
RAB41
Gene Name
RAB41, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details