Product Details

SNP ID
rs201574982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:54540230 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATGCCAATCGAGAGGCTGAATT[A/G]AAGAAGAAGTGGGTAAGCTTTTTGC
Phenotype
MIM: 300873
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNL3L PubMed Links

Gene Details

Gene
GNL3L
Gene Name
G protein nucleolar 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184819.1 316 Silent Mutation TTA,TTG L59L NP_001171748.1
NM_019067.5 316 Silent Mutation TTA,TTG L59L NP_061940.1

View Full Product Details