Product Details
- SNP ID
-
rs201369085
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:134807614 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGACCCGGACATGGCACAGGAGAAA[A/G]TGAAACTAGGTTTCAAGTCGCTGCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FAM122C
PubMed Links
Gene Details
- Gene
- FAM122C
- Gene Name
- family with sequence similarity 122C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001170779.1 |
288 |
Missense Mutation |
ATG,GTG |
M6V |
NP_001164250.1 |
NM_001170780.1 |
288 |
Missense Mutation |
ATG,GTG |
M42V |
NP_001164251.1 |
NM_001170781.1 |
288 |
Missense Mutation |
ATG,GTG |
M6V |
NP_001164252.1 |
NM_001170782.1 |
288 |
Missense Mutation |
ATG,GTG |
M6V |
NP_001164253.1 |
NM_001170783.1 |
288 |
Missense Mutation |
ATG,GTG |
M6V |
NP_001164254.1 |
NM_001170784.1 |
288 |
Missense Mutation |
ATG,GTG |
M6V |
NP_001164255.1 |
NM_138819.3 |
288 |
Missense Mutation |
ATG,GTG |
M6V |
NP_620174.1 |
XM_005262382.1 |
288 |
Missense Mutation |
ATG,GTG |
M42V |
XP_005262439.1 |
XM_005262383.2 |
288 |
Missense Mutation |
ATG,GTG |
M42V |
XP_005262440.1 |
XM_005262384.4 |
288 |
Missense Mutation |
ATG,GTG |
M42V |
XP_005262441.1 |
XM_005262386.3 |
288 |
Missense Mutation |
ATG,GTG |
M42V |
XP_005262443.1 |
XM_005262387.3 |
288 |
Missense Mutation |
ATG,GTG |
M42V |
XP_005262444.1 |
XM_006724734.3 |
288 |
Missense Mutation |
ATG,GTG |
M6V |
XP_006724797.1 |
XM_006724735.1 |
288 |
Missense Mutation |
ATG,GTG |
M42V |
XP_006724798.1 |
XM_006724736.2 |
288 |
Missense Mutation |
ATG,GTG |
M42V |
XP_006724799.1 |
XM_011531295.1 |
288 |
Missense Mutation |
ATG,GTG |
M42V |
XP_011529597.1 |
XM_011531296.2 |
288 |
Missense Mutation |
ATG,GTG |
M42V |
XP_011529598.1 |
XM_011531297.2 |
288 |
Missense Mutation |
ATG,GTG |
M42V |
XP_011529599.1 |
XM_011531298.2 |
288 |
Missense Mutation |
ATG,GTG |
M42V |
XP_011529600.1 |
XM_011531299.1 |
288 |
Intron |
|
|
XP_011529601.1 |
XM_011531300.2 |
288 |
Intron |
|
|
XP_011529602.1 |
XM_011531301.2 |
288 |
Intron |
|
|
XP_011529603.1 |
XM_011531302.2 |
288 |
Intron |
|
|
XP_011529604.1 |
XM_011531305.2 |
288 |
Missense Mutation |
ATG,GTG |
M42V |
XP_011529607.1 |
XM_011531306.2 |
288 |
Missense Mutation |
ATG,GTG |
M42V |
XP_011529608.1 |
XM_017029308.1 |
288 |
Missense Mutation |
ATG,GTG |
M42V |
XP_016884797.1 |
XM_017029309.1 |
288 |
Intron |
|
|
XP_016884798.1 |
XM_017029310.1 |
288 |
Missense Mutation |
ATG,GTG |
M42V |
XP_016884799.1 |
XM_017029311.1 |
288 |
Missense Mutation |
ATG,GTG |
M6V |
XP_016884800.1 |
View Full Product Details