Product Details

SNP ID
rs6014946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:57169255 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAACTGTCTGAGTCAAGATGGAGA[A/C]ACAGTCCCCCACAAGACCGGGTTCC
Phenotype
MIM: 112267
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BMP7 PubMed Links

Gene Details

Gene
BMP7
Gene Name
bone morphogenetic protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001719.2 3529 UTR 3 NP_001710.1

View Full Product Details