Product Details

SNP ID
rs6719211
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:45389962 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCACAAACACTGAGAAAGTAAGACC[G/T]TTGGAAACAAGAGACTGGAAGCTTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SRBD1 PubMed Links

Gene Details

Gene
SRBD1
Gene Name
S1 RNA binding domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018079.4 Intron NP_060549.4
XM_011532946.1 Intron XP_011531248.1
XM_017004417.1 Intron XP_016859906.1
XM_017004418.1 Intron XP_016859907.1

View Full Product Details