Product Details

SNP ID
rs11064860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110051403 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTATCTGGCACATACTGAGCACT[C/T]GCTATAGGAAAGCTGCTGCCCCCAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf76 PubMed Links

Gene Details

Gene
C12orf76
Gene Name
chromosome 12 open reading frame 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207435.2 Intron NP_997318.1
XM_017019297.1 Intron XP_016874786.1

View Full Product Details