Product Details

SNP ID
rs8005382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:71192365 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGAGGAAAAGTGAGGAATCTTCTG[C/T]GGCCCCAGTGTTGAGAATGAGGTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105370706 PubMed Links

Gene Details

Gene
LOC105370706
Gene Name
uncharacterized LOC105370706
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011537452.2 Intron XP_011535754.2

View Full Product Details