Product Details
- SNP ID
-
rs2062925
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:9634554 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CGTCTAGAAGGTTAGTCCTGAAATT[G/T]TGTGTGTGTATGTGTGTGTACATAC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PER4
PubMed Links
Gene Details
- Gene
- PER4
- Gene Name
- period circadian clock 3 pseudogene
There are no transcripts associated with this gene.
View Full Product Details