Product Details

SNP ID
rs5957383
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:120430837 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAAAACCAAGTTTAAGCATATCTGA[C/T]GGGCATTTATACCACATCAATATTT
Phenotype
MIM: 309060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAMP2 PubMed Links

Gene Details

Gene
LAMP2
Gene Name
lysosomal associated membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122606.1 1899 Intron NP_001116078.1
NM_002294.2 1899 UTR 3 NP_002285.1
NM_013995.2 1899 Intron NP_054701.1
Gene
LOC105373326
Gene Name
uncharacterized LOC105373326
There are no transcripts associated with this gene.

View Full Product Details