Product Details

SNP ID
rs12689903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103630721 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTATATTTATATATTAATATGCA[A/T]AGAAAAAGCTAAAAGTATAGACTTC
Phenotype
MIM: 300237
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TCEAL1 PubMed Links

Gene Details

Gene
TCEAL1
Gene Name
transcription elongation factor A like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006639.1 967 UTR 3 NP_001006640.1
NM_001006640.1 967 UTR 3 NP_001006641.1
NM_004780.2 967 UTR 3 NP_004771.2

View Full Product Details