Product Details

SNP ID
rs5712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20770008 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGAACTGGTCTCTGTGCAAATCC[C/T]GAGTGCTAAAGCTTCCAACAAGACT
Phenotype
MIM: 145505
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACSM3 PubMed Links

Gene Details

Gene
ACSM3
Gene Name
acyl-CoA synthetase medium-chain family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005622.3 155 UTR 5 NP_005613.2
NM_202000.2 155 UTR 5 NP_973729.1
XM_011545911.1 155 UTR 5 XP_011544213.1
XM_017023523.1 155 UTR 5 XP_016879012.1
XM_017023524.1 155 UTR 5 XP_016879013.1

View Full Product Details