Product Details

SNP ID
rs6104293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45697659 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCCACCACACCTAGCTGACATCC[C/T]ATATTCTATATTGGAAGACTTAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR3617 PubMed Links

Gene Details

Gene
MIR3617
Gene Name
microRNA 3617
There are no transcripts associated with this gene.

Gene
WFDC10B
Gene Name
WAP four-disulfide core domain 10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172006.2 Intron NP_742003.1
NM_172131.2 Intron NP_742143.1
XM_017027824.1 Intron XP_016883313.1
XM_017027825.1 Intron XP_016883314.1
Gene
WFDC13
Gene Name
WAP four-disulfide core domain 13
There are no transcripts associated with this gene.

View Full Product Details