Product Details

SNP ID
rs35885527
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:57759439 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTACTGCACAGGCTCAGCCAGCCCA[C/T]GCTCAGAGAGCAGGGTCCCAGGGTA
Phenotype
MIM: 602703 MIM: 604535
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KATNB1 PubMed Links

Gene Details

Gene
KATNB1
Gene Name
katanin regulatory subunit B1
There are no transcripts associated with this gene.

Gene
KIFC3
Gene Name
kinesin family member C3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130099.1 Intron NP_001123571.1
NM_001130100.1 Intron NP_001123572.1
NM_001318710.1 Intron NP_001305639.1
NM_001318711.1 Intron NP_001305640.1
NM_001318712.1 Intron NP_001305641.1
NM_001318713.1 Intron NP_001305642.1
NM_001318714.1 Intron NP_001305643.1
NM_001318715.1 Intron NP_001305644.1
NM_005550.3 Intron NP_005541.3
XM_005255937.1 Intron XP_005255994.1
XM_005255944.1 Intron XP_005256001.1
XM_006721188.1 Intron XP_006721251.1
XM_011523075.1 Intron XP_011521377.1
XM_011523076.1 Intron XP_011521378.1
XM_011523077.1 Intron XP_011521379.1
XM_011523078.1 Intron XP_011521380.1
XM_011523079.1 Intron XP_011521381.1
XM_017023219.1 Intron XP_016878708.1
XM_017023220.1 Intron XP_016878709.1
XM_017023221.1 Intron XP_016878710.1
XM_017023222.1 Intron XP_016878711.1
XM_017023223.1 Intron XP_016878712.1
XM_017023224.1 Intron XP_016878713.1
XM_017023225.1 Intron XP_016878714.1
XM_017023226.1 Intron XP_016878715.1
XM_017023227.1 Intron XP_016878716.1

View Full Product Details