Product Details

SNP ID
rs10786352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97511524 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGCCAATTCTTGCACTATATATC[C/T]GAATGTGCCAGAAAGTGCACTTTGT
Phenotype
MIM: 616388
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBTD1 PubMed Links

Gene Details

Gene
UBTD1
Gene Name
ubiquitin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024954.4 Intron NP_079230.1

View Full Product Details