Product Details

SNP ID
rs12571363
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:122471096 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACGCTCCTCCTGGGGTGGCCTGCAT[A/C]CTGTGTTCTTCTCAGGCCGACTTTC
Phenotype
MIM: 602194
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HTRA1 PubMed Links

Gene Details

Gene
HTRA1
Gene Name
HtrA serine peptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002775.4 Intron NP_002766.1

View Full Product Details