Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001201465.1 | 2052 | Intron | NP_001188394.1 | ||
NM_138966.3 | 2052 | Intron | NP_620416.1 | ||
NM_138999.1 | 2052 | Intron | NP_620552.1 | ||
XM_005266774.4 | 2052 | Intron | XP_005266831.1 | ||
XM_017026017.1 | 2052 | UTR 3 | XP_016881506.1 | ||
XM_017026018.1 | 2052 | Intron | XP_016881507.1 | ||
XM_017026019.1 | 2052 | UTR 3 | XP_016881508.1 | ||
XM_017026020.1 | 2052 | Intron | XP_016881509.1 | ||
XM_017026021.1 | 2052 | Intron | XP_016881510.1 | ||
XM_017026022.1 | 2052 | Intron | XP_016881511.1 | ||
XM_017026023.1 | 2052 | Intron | XP_016881512.1 | ||
XM_017026024.1 | 2052 | Intron | XP_016881513.1 |