Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_052846.1 | 3687 | UTR 3 | NP_443078.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001301860.1 | 3687 | UTR 3 | NP_001288789.1 | ||
XM_005260516.2 | 3687 | Intron | XP_005260573.1 | ||
XM_006723863.2 | 3687 | Intron | XP_006723926.1 | ||
XM_011528996.1 | 3687 | UTR 3 | XP_011527298.1 | ||
XM_011528997.2 | 3687 | UTR 3 | XP_011527299.1 | ||
XM_011528998.2 | 3687 | UTR 3 | XP_011527300.1 | ||
XM_011528999.2 | 3687 | UTR 3 | XP_011527301.1 | ||
XM_011529000.1 | 3687 | UTR 3 | XP_011527302.1 | ||
XM_011529001.1 | 3687 | UTR 3 | XP_011527303.1 | ||
XM_011529002.2 | 3687 | UTR 3 | XP_011527304.1 | ||
XM_011529003.1 | 3687 | Intron | XP_011527305.1 | ||
XM_011529004.1 | 3687 | Intron | XP_011527306.1 | ||
XM_011529005.2 | 3687 | Intron | XP_011527307.1 | ||
XM_011529006.2 | 3687 | Intron | XP_011527308.1 | ||
XM_017028020.1 | 3687 | UTR 3 | XP_016883509.1 |