Product Details

SNP ID
rs6129731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:41032047 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTGATTTATGTAACTATGAAAAA[A/C]ACTTTAAAAAAAGTAGAAATCAATG
Phenotype
MIM: 126420
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC100128988 PubMed Links

Gene Details

Gene
LOC100128988
Gene Name
uncharacterized LOC100128988
There are no transcripts associated with this gene.

Gene
TOP1
Gene Name
topoisomerase (DNA) I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003286.2 Intron NP_003277.1

View Full Product Details