Product Details

SNP ID
rs7776426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:129831375 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCACCAATGTGATGTCAAGGGGA[A/C]TTCCAACATAACTGCAATAGAAAAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMEM244 PubMed Links

Gene Details

Gene
TMEM244
Gene Name
transmembrane protein 244
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010876.1 414 Missense Mutation GTC,TTC V111F NP_001010876.1
XM_011535669.2 414 Intron XP_011533971.1

View Full Product Details