Product Details

SNP ID
rs34871527
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3046446 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGTTCTGGCGGAGCGCAAGGCC[C/T]TGAAGGACGCCGCCGAGCACCGCGA
Phenotype
MIM: 602352 MIM: 611988 MIM: 176884
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GNRH2 PubMed Links

Gene Details

Gene
GNRH2
Gene Name
gonadotropin releasing hormone 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310220.1 326 Intron NP_001297149.1
NM_001501.1 326 Intron NP_001492.1
NM_178331.1 326 Intron NP_847901.1
NM_178332.1 326 Intron NP_847902.1
XM_017027823.1 326 Intron XP_016883312.1
Gene
MRPS26
Gene Name
mitochondrial ribosomal protein S26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030811.3 326 Silent Mutation CTG,TTG L96L NP_110438.1
Gene
PTPRA
Gene Name
protein tyrosine phosphatase, receptor type A
There are no transcripts associated with this gene.

View Full Product Details