Product Details

SNP ID
rs34601747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56984176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGCGCCCAGCCTGAAAAAAAAT[A/C]TGATAAATGGATGAGTAAATGAGTC
Phenotype
MIM: 614919 MIM: 180661
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NOA1 PubMed Links

Gene Details

Gene
NOA1
Gene Name
nitric oxide associated 1
There are no transcripts associated with this gene.

Gene
POLR2B
Gene Name
RNA polymerase II subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000938.2 Intron NP_000929.1
NM_001303268.1 Intron NP_001290197.1
NM_001303269.1 Intron NP_001290198.1

View Full Product Details