Product Details

SNP ID
rs9998265
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:70383539 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATAGTCTCAGGTGTACTCTAAAAG[A/T]CAGAATTGCTCTACAGAATATTTTT
Phenotype
MIM: 611593
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SMR3B PubMed Links

Gene Details

Gene
SMR3B
Gene Name
submaxillary gland androgen regulated protein 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006685.3 Intron NP_006676.1

View Full Product Details