Product Details

SNP ID
rs6585808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122276626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATCCTTGGTCTATGGAATACAG[C/T]GTCTTTGCCTTGAGTTGTCTTGTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTBD16 PubMed Links

Gene Details

Gene
BTBD16
Gene Name
BTB domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318189.1 Intron NP_001305118.1
NM_144587.3 Intron NP_653188.2
XM_011539239.2 Intron XP_011537541.1
XM_011539240.2 Intron XP_011537542.1
XM_011539241.2 Intron XP_011537543.1
XM_011539242.2 Intron XP_011537544.1
XM_017015636.1 Intron XP_016871125.1
XM_017015637.1 Intron XP_016871126.1

View Full Product Details