Product Details

SNP ID
rs2438113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:103848552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAATTGTAAAGAGGAAGGAAGG[C/T]CTGGCACTATCTAAAGGCTCTTTGT
Phenotype
MIM: 611076
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NT5DC3 PubMed Links

Gene Details

Gene
NT5DC3
Gene Name
5'-nucleotidase domain containing 3
There are no transcripts associated with this gene.

Gene
TTC41P
Gene Name
tetratricopeptide repeat domain 41, pseudogene
There are no transcripts associated with this gene.

View Full Product Details