Product Details

SNP ID
rs11744596
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:69223464 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCAGCATTTTCTCACAGCCACAGAA[G/T]TACAGTGGTGAATTTTTAGGCCAAT
Phenotype
MIM: 611996
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MRPS36 PubMed Links

Gene Details

Gene
MRPS36
Gene Name
mitochondrial ribosomal protein S36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033281.5 Intron NP_150597.1

View Full Product Details