Product Details

SNP ID
rs2972381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:69242415 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACCAGTTGAGTCAGTTTCTTCATA[C/T]GGGCTCCTGGTCCAAGTGGCACCAG
Phenotype
MIM: 601955
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK7 PubMed Links

Gene Details

Gene
CDK7
Gene Name
cyclin dependent kinase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324069.1 Intron NP_001310998.1
NM_001324070.1 Intron NP_001310999.1
NM_001324071.1 Intron NP_001311000.1
NM_001324072.1 Intron NP_001311001.1
NM_001324074.1 Intron NP_001311003.1
NM_001324075.1 Intron NP_001311004.1
NM_001324077.1 Intron NP_001311006.1
NM_001324078.1 Intron NP_001311007.1
NM_001799.3 Intron NP_001790.1
XM_011543094.2 Intron XP_011541396.1

View Full Product Details