Product Details

SNP ID
rs17318799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:26519697 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAACTAATATTTCCTAATCTAGTA[C/T]CGTATTTCTTCCAATAGTCAATTAC
Phenotype
MIM: 602463 MIM: 603970
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPYSL2 PubMed Links

Gene Details

Gene
DPYSL2
Gene Name
dihydropyrimidinase like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197293.2 Intron NP_001184222.1
NM_001244604.1 Intron NP_001231533.1
NM_001386.5 Intron NP_001377.1
Gene
PNMA2
Gene Name
paraneoplastic Ma antigen 2
There are no transcripts associated with this gene.

View Full Product Details