Product Details

SNP ID
rs6879867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:65151833 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGGCACTCTCTCTGGCTTTCTGT[A/G]GGTCAGTGTCCTTGGCAGGTCTTAC
Phenotype
MIM: 605008
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS6 PubMed Links

Gene Details

Gene
ADAMTS6
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_197941.3 2595 UTR 3 NP_922932.2
XM_011543113.2 2595 UTR 3 XP_011541415.1
XM_011543114.2 2595 UTR 3 XP_011541416.1
XM_011543115.2 2595 UTR 3 XP_011541417.1
XM_011543116.2 2595 UTR 3 XP_011541418.1
XM_011543117.1 2595 UTR 3 XP_011541419.1
XM_011543119.2 2595 Intron XP_011541421.1
XM_011543120.2 2595 Intron XP_011541422.1
XM_011543121.2 2595 UTR 3 XP_011541423.2
XM_011543122.2 2595 Intron XP_011541424.1
XM_011543123.2 2595 UTR 3 XP_011541425.1
XM_011543124.2 2595 UTR 3 XP_011541426.1
XM_011543125.1 2595 UTR 3 XP_011541427.1
XM_011543126.2 2595 Intron XP_011541428.1
XM_011543127.2 2595 Intron XP_011541429.1

View Full Product Details