Product Details
- SNP ID
-
rs12667647
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:1686115 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CCTGTTTCATTGGATGTCTTGTAAG[C/T]TTGTGTTGAAAACAAACACTTCAGG
- Phenotype
-
MIM: 614964
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ELFN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115533354] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ELFN1
- Gene Name
- extracellular leucine rich repeat and fibronectin type III domain containing 1
- Gene
- LOC401296
- Gene Name
- uncharacterized LOC401296
There are no transcripts associated with this gene.
View Full Product Details