Product Details

SNP ID
rs11716540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:128736890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTAGTATCATACTTGGCTTATAA[C/T]ATGTAATAGCTATTGGGGGAAATAG
Phenotype
MIM: 602298
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB7A PubMed Links

Gene Details

Gene
RAB7A
Gene Name
RAB7A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004637.5 Intron NP_004628.4

View Full Product Details