Product Details

SNP ID
rs2981013
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:128469500 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGTTGCTTCAGCCAGTTAGGGCA[A/G]CTCCCCTCCTCCAGGGTCCCAGCAC
Phenotype
MIM: 611337 MIM: 137295
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJB8 PubMed Links

Gene Details

Gene
DNAJB8
Gene Name
DnaJ heat shock protein family (Hsp40) member B8
There are no transcripts associated with this gene.

Gene
DNAJB8-AS1
Gene Name
DNAJB8 antisense RNA 1
There are no transcripts associated with this gene.

Gene
GATA2
Gene Name
GATA binding protein 2
There are no transcripts associated with this gene.

View Full Product Details