Product Details
- SNP ID
-
rs7638833
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:125978329 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTTTTTCACACCACATTTGGAGA[C/T]GTTTTGGGGGAAAATGTACCTTTAA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALG1L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs149297214] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALG1L
- Gene Name
- ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase like
- Gene
- ROPN1B
- Gene Name
- rhophilin associated tail protein 1B
View Full Product Details