Product Details

SNP ID
rs3749598
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:161386160 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGAACCCTTACAAGACCTTTCACAT[A/C]ATTAATGCTGACAAGGTAGTGGCCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FSTL5 PubMed Links

Gene Details

Gene
FSTL5
Gene Name
follistatin like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128427.2 2564 Missense Mutation NP_001121899.1
NM_001128428.2 2564 Missense Mutation NP_001121900.1
NM_020116.4 2564 Missense Mutation NP_064501.2
XM_011532126.1 2564 Missense Mutation XP_011530428.1

View Full Product Details