Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001166108.1 | 78 | UTR 5 | NP_001159580.1 | ||
NM_001166109.1 | 78 | Intron | NP_001159581.1 | ||
NM_001166110.1 | 78 | Intron | NP_001159582.1 | ||
NM_016081.3 | 78 | UTR 5 | NP_057165.3 | ||
XM_005262861.4 | 78 | UTR 5 | XP_005262918.1 | ||
XM_005262866.2 | 78 | Intron | XP_005262923.1 | ||
XM_011531768.2 | 78 | Intron | XP_011530070.1 | ||
XM_011531769.2 | 78 | Intron | XP_011530071.1 | ||
XM_011531770.2 | 78 | Intron | XP_011530072.1 | ||
XM_011531771.2 | 78 | Intron | XP_011530073.1 | ||
XM_011531772.2 | 78 | Intron | XP_011530074.1 | ||
XM_011531773.1 | 78 | Intron | XP_011530075.1 | ||
XM_011531774.1 | 78 | Intron | XP_011530076.1 | ||
XM_011531775.1 | 78 | Intron | XP_011530077.1 | ||
XM_017007910.1 | 78 | Intron | XP_016863399.1 | ||
XM_017007911.1 | 78 | Intron | XP_016863400.1 |