Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001197246.2 | Intron | NP_001184175.1 | |||
NM_001197247.2 | Intron | NP_001184176.1 | |||
NM_001197248.2 | Intron | NP_001184177.1 | |||
NM_001197249.2 | Intron | NP_001184178.1 | |||
NM_007047.4 | Intron | NP_008978.2 | |||
XM_005248827.3 | Intron | XP_005248884.1 | |||
XM_005248831.3 | Intron | XP_005248888.1 | |||
XM_005248832.3 | Intron | XP_005248889.1 | |||
XM_006714979.3 | Intron | XP_006715042.1 | |||
XM_006714980.3 | Intron | XP_006715043.1 | |||
XM_006714981.3 | Intron | XP_006715044.1 | |||
XM_006714982.3 | Intron | XP_006715045.1 | |||
XM_011514267.2 | Intron | XP_011512569.1 | |||
XM_011514268.2 | Intron | XP_011512570.1 | |||
XM_011514269.2 | Intron | XP_011512571.1 | |||
XM_011514270.2 | Intron | XP_011512572.1 | |||
XM_011514271.2 | Intron | XP_011512573.1 | |||
XM_017010211.1 | Intron | XP_016865700.1 | |||
XM_017010212.1 | Intron | XP_016865701.1 | |||
XM_017010213.1 | Intron | XP_016865702.1 | |||
XM_017010214.1 | Intron | XP_016865703.1 | |||
XM_017010215.1 | Intron | XP_016865704.1 | |||
XM_017010216.1 | Intron | XP_016865705.1 | |||
XM_017010217.1 | Intron | XP_016865706.1 |