Product Details

SNP ID
rs12990344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:94773260 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTGTTTGCAGAATGGTGCTCCCTG[G/T]GTGGGGATTCCAACCCTATATATTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD20A8P PubMed Links

Gene Details

Gene
ANKRD20A8P
Gene Name
ankyrin repeat domain 20 family member A8, pseudogene
There are no transcripts associated with this gene.

View Full Product Details