Product Details

SNP ID
rs5018567
Assay Type
Functionally tested
NCBI dbSNP Submissions
60
Location
Chr.1:154507842 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTCCCAAATGTGCTCAGCTTTTT[C/T]GTATCTCCATGTCCTGGCATGGGCT
Phenotype
MIM: 610482
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SHE PubMed Links

Gene Details

Gene
SHE
Gene Name
Src homology 2 domain containing E
There are no transcripts associated with this gene.

Gene
TDRD10
Gene Name
tudor domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098475.1 Intron NP_001091945.1
NM_182499.3 Intron NP_872305.3
XM_006711156.2 Intron XP_006711219.1
XM_011509152.1 Intron XP_011507454.1
XM_011509153.1 Intron XP_011507455.1
XM_011509154.1 Intron XP_011507456.1
XM_011509155.1 Intron XP_011507457.1
XM_011509156.1 Intron XP_011507458.1
XM_011509157.1 Intron XP_011507459.1
XM_011509158.2 Intron XP_011507460.1
XM_011509159.2 Intron XP_011507461.1
XM_011509160.2 Intron XP_011507462.1
XM_011509161.1 Intron XP_011507463.1
XM_011509162.2 Intron XP_011507464.1
XM_017000255.1 Intron XP_016855744.1

View Full Product Details