Product Details
- SNP ID
-
rs11800158
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
13
- Location
-
Chr.1:155084386 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGGGCAGGGGGAGACCCCCAGAT[C/T]GGTGGGGTCAACCAATACTGGCATG
- Phenotype
-
MIM: 601381
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EFNA3
PubMed Links
Gene Details
- Gene
- EFNA3
- Gene Name
- ephrin A3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004952.4 |
|
Intron |
|
|
NP_004943.1 |
View Full Product Details