Product Details
- SNP ID
-
rs4515830
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
25
- Location
-
Chr.1:153663996 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTACTACTACTACTACTACTACTAC[C/T]AGTAAATAAGGATGATGAGGAACTC
- Phenotype
-
MIM: 603181
MIM: 607007
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ILF2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77282452] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ILF2
- Gene Name
- interleukin enhancer binding factor 2
- Gene
- SNAPIN
- Gene Name
- SNAP associated protein
There are no transcripts associated with this gene.
View Full Product Details