Product Details

SNP ID
rs567969
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:173008013 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGCTACAAGATCAAAGAGGTACCAC[A/G]TTCTGGCCTCGAGCCATAGTAGGCT
Phenotype
MIM: 603931
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6V0E1 PubMed Links

Gene Details

Gene
ATP6V0E1
Gene Name
ATPase H+ transporting V0 subunit e1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003945.3 Intron NP_003936.1

View Full Product Details