Product Details

SNP ID
rs678849
Assay Type
Validated
NCBI dbSNP Submissions
67
Location
Chr.1:28818676 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
GTCCTTCTTACCATAGTGTCAAAAG[C/T]ACCTGCTAGGTGCTGAGCTTGGCTG
Phenotype
MIM: 165195
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OPRD1 PubMed Links

Gene Details

Gene
OPRD1
Gene Name
opioid receptor delta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000911.3 Intron NP_000902.3

View Full Product Details