Product Details

SNP ID
rs12626098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62645983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTGCCACTGGACTCTGCCTTTC[A/G]GGAGGCGCTGTGGACCTGAGATTTG
Phenotype
MIM: 612436
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLCO4A1 PubMed Links

Gene Details

Gene
SLCO4A1
Gene Name
solute carrier organic anion transporter family member 4A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016354.3 Intron NP_057438.3
XM_005260203.2 Intron XP_005260260.1
XM_011528792.2 Intron XP_011527094.1
XM_017027826.1 Intron XP_016883315.1
XM_017027827.1 Intron XP_016883316.1

View Full Product Details