Product Details

SNP ID
rs2488238
Assay Type
Functionally tested
NCBI dbSNP Submissions
67
Location
Chr.1:30872105 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGTCTGACACTGGCTGGGGTTGG[C/T]GGCTGCATGGTTGGTGTGGGCACCA
Phenotype
MIM: 186357
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC400748 PubMed Links

Gene Details

Gene
LOC400748
Gene Name
uncharacterized LOC400748
There are no transcripts associated with this gene.

Gene
SDC3
Gene Name
syndecan 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014654.3 2482 UTR 3 NP_055469.3
XM_011542463.1 2482 UTR 3 XP_011540765.1
XM_011542464.2 2482 UTR 3 XP_011540766.1
XM_011542466.1 2482 UTR 3 XP_011540768.1

View Full Product Details