Product Details

SNP ID
rs12566400
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:36163065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGAGTTGTTGGGGTGGGAGGAG[A/G]GAGAGACAAAAGGAAAGGACTGCCA
Phenotype
MIM: 610955
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP7D1 PubMed Links

Gene Details

Gene
MAP7D1
Gene Name
MAP7 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286365.1 Intron NP_001273294.1
NM_001286366.1 Intron NP_001273295.1
NM_018067.4 Intron NP_060537.3
XM_005271027.1 Intron XP_005271084.1
XM_006710759.1 Intron XP_006710822.1
XM_006710761.1 Intron XP_006710824.1
Gene
TRAPPC3
Gene Name
trafficking protein particle complex 3
There are no transcripts associated with this gene.

View Full Product Details