Product Details

SNP ID
rs12656542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150932060 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGCAGTCATGAAATATTTTTCCA[A/G]ATATATTATATGGATGACCTGACTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF300P1 PubMed Links

Gene Details

Gene
ZNF300P1
Gene Name
zinc finger protein 300 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details