Product Details

SNP ID
rs7978610
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:123984025 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAGTGTGAAGAGCCGAGGTTTCTGA[C/G]CAGGTGTTTGAGTGCCCTCTGGAGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF664 PubMed Links

Gene Details

Gene
ZNF664
Gene Name
zinc finger protein 664
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204298.1 Intron NP_001191227.1
NM_152437.2 Intron NP_689650.1
Gene
ZNF664-FAM101A
Gene Name
filamin-interacting protein FAM101A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204299.1 Intron NP_001191228.1

View Full Product Details