Product Details

SNP ID
rs12515110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176811547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGCCTTTCTTGGCCCCTGCATGC[T/C]CCATTGGTCACCCTAACCCACTGCT
Phenotype
MIM: 607869
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
UNC5A PubMed Links

Gene Details

Gene
UNC5A
Gene Name
unc-5 netrin receptor A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133369.2 Intron NP_588610.2
XM_006714927.1 Intron XP_006714990.1
XM_006714928.1 Intron XP_006714991.1
XM_011534686.1 Intron XP_011532988.1
XM_011534687.2 Intron XP_011532989.1

View Full Product Details