Product Details

SNP ID
rs35051373
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:5714520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTTTTTTTTTTTTTTTGGCTGC[A/G]TGCAATAGCCGTTGACCCTCCACTG
Phenotype
MIM: 604831 MIM: 607261
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
EVC PubMed Links
Additional Information
For this assay, SNP(s) [rs71171414] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
EVC
Gene Name
EvC ciliary complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306090.1 Intron NP_001293019.1
NM_001306092.1 Intron NP_001293021.1
NM_153717.2 Intron NP_714928.1
XM_006713865.3 Intron XP_006713928.1
XM_006713866.3 Intron XP_006713929.1
XM_011513419.2 Intron XP_011511721.1
XM_017007883.1 Intron XP_016863372.1
Gene
EVC2
Gene Name
EvC ciliary complex subunit 2
There are no transcripts associated with this gene.

View Full Product Details